Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpte | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199257.2 | NP_954866.2 | 26, 27 | 2202 | ||
GenBank mRNA | AJ311311.1 | - | 26, 27 | 2205 | ||
AJ311312.1 | - | 26, 27 | 2290 | |||
AJ311313.1 | - | 27, 28 | 2328 | |||
BC107329.2 | - | 26, 27 | 1997 | 40054415 |