Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cib3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080812.1 | NP_001074281.1 | 2 | 156 | ||
GenBank mRNA | AK050094.1 | - | 2 | 156 | ||
BB870365.1 | - | 2 | 181 | |||
BC120812.1 | - | 2 | 138 | 40129735 | ||
BC120814.1 | - | 2 | 138 | 40129737 |