Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024617.3 | NP_001019788.1 | 11 | 1165 | ||
NM_001297591.1 | NP_001284520.1 | 11 | 1142 | |||
NM_001297593.1 | NP_001284522.1 | 10 | 1023 | |||
NM_001297596.1 | NP_001284525.1 | 11 | 1142 | |||
GenBank mRNA | AK053554.1 | - | 4 | 294 | ||
AK089068.1 | - | 6 | 396 | |||
AK139397.1 | - | 4 | 643 | |||
AK141502.1 | - | 13 | 1139 | |||
BC064813.1 | - | 9 | 867 | 6826283 | ||
DQ296611.1 | - | 11 | 1142 | |||
DQ296612.1 | - | 10 | 1023 | |||
DQ296613.1 | - | 11 | 1142 |