Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vac14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146216.2 | NP_666328.2 | 11 | 1456 | ||
GenBank mRNA | AK031952.1 | - | 11 | 1491 | ||
AK043319.1 | - | 11 | 1488 | |||
AK046826.1 | - | 11 | 1428 | |||
AK054468.1 | - | 2 | 283 | |||
AK084520.1 | - | 11 | 1507 | |||
AK140191.1 | - | 11 | 1483 | |||
AK171577.1 | - | 11 | 1496 | |||
AK171883.1 | - | 11 | 1484 | |||
AK183924.1 | - | 2 | 116 | |||
AK185154.1 | - | 3 | 268 | |||
AK187571.1 | - | 3 | 268 | |||
AK190879.1 | - | 3 | 290 | |||
AK197455.1 | - | 3 | 268 | |||
AK198087.1 | - | 3 | 268 | |||
AK199043.1 | - | 4 | 269 | |||
AK202260.1 | - | 3 | 268 | |||
AK203281.1 | - | 3 | 270 | |||
AK203406.1 | - | 3 | 267 | |||
AK203450.1 | - | 3 | 268 | |||
AK206758.1 | - | 3 | 284 | |||
AK209742.1 | - | 3 | 268 | |||
AK211234.1 | - | 3 | 268 | |||
AK211787.1 | - | 3 | 288 | |||
BC032215.1 | - | 4 | 369 | |||
BC052199.1 | - | 11 | 1456 | 6512678 |