Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atmin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177700.4 | NP_808368.3 | 4 | 1787 | ||
GenBank mRNA | AK087334.1 | - | 1 | 1098 | ||
AK122281.1 | - | 4 | 1669 | |||
AX775769.1 | - | 4 | 1787 | |||
BC025922.1 | - | 1 | 393 | |||
BC058974.1 | - | 3 | 1332 | |||
BC060631.1 | - | 4 | 1772 | 6847850 |