Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145607.3 | NP_663582.2 | 16, 17 | 1892 | ||
GenBank mRNA | AK144842.1 | - | 16, 17 | 1893 | ||
AK148102.1 | - | 18, 19 | 2122 | |||
AK160894.1 | - | 18, 19 | 2129 | |||
BC017545.1 | - | 10, 11 | 1103 | |||
BC072577.1 | - | 16, 17 | 1882 | |||
BC079851.1 | - | 17, 18 | 1947 | 6401930 |