Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205367.1 | NP_001192296.1 | 10 | 1107 | ||
NM_009859.4 | NP_033989.2 | 11 | 1110 | |||
GenBank mRNA | AK052529.1 | - | 2 | 246 | ||
AK088997.1 | - | 11 | 897 | |||
AK144127.1 | - | 11 | 899 | |||
AK220447.1 | - | 10 | 880 | |||
BC058587.1 | - | 10 | 911 | 6529169 |