Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004805.3 | NP_004796.1 | 1, 2 | 123 | ||
GenBank mRNA | BC017205.2 | - | 1, 2 | 95 | 3617979 | |
BC093795.1 | - | 1, 2 | 123 | 7939640 | ||
BC093797.1 | - | 1, 2 | 125 | 7939642 | ||
U85510.1 | - | 1, 2 | 91 |