Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291448.1 | NP_001278377.1 | 7, 8 | 969 | ||
NM_001291449.1 | NP_001278378.1 | 7, 8 | 974 | |||
NM_172291.2 | NP_758495.1 | 7, 8 | 969 | |||
GenBank mRNA | AK054231.1 | - | 7 | 861 | ||
AK137070.1 | - | 1 | 460 | |||
AK144759.1 | - | 7, 8 | 895 | |||
AK163722.1 | - | 7, 8 | 828 | |||
BC017604.1 | - | 7, 8 | 927 | |||
BC024806.1 | - | 7, 8 | 839 | 5361053 | ||
BC027386.1 | - | 4, 5 | 1448 | |||
BC028892.1 | - | 5, 6 | 1940 |