Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apeh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146226.2 | NP_666338.2 | 14 | 1283 | ||
GenBank mRNA | AK041271.1 | - | 10 | 3091 | ||
AK149671.1 | - | 14 | 1287 | |||
AK149713.1 | - | 14 | 1285 | |||
AK217552.1 | - | 1 | 30 | |||
AK218199.1 | - | 4 | 260 | |||
BC005716.1 | - | 13 | 1409 | |||
BC023937.1 | - | 4 | 252 | |||
BC025494.1 | - | 14 | 1265 | 5310043 | ||
BC026594.1 | - | 9 | 816 |