Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apeh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146226.2 | NP_666338.2 | 10, 11 | 1039 | ||
GenBank mRNA | AK149671.1 | - | 10, 11 | 1044 | ||
AK149713.1 | - | 10, 11 | 1041 | |||
AK179741.1 | - | 1, 2 | 142 | |||
AK218199.1 | - | 1 | 16 | |||
BC005716.1 | - | 9, 10 | 1165 | |||
BC025494.1 | - | 10, 11 | 1021 | 5310043 | ||
BC026594.1 | - | 5, 6 | 572 |