Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081340.2 | NP_001074809.2 | 17 | 7172 | ||
GenBank mRNA | AK009337.1 | - | 3 | 633 | ||
AK134444.1 | - | 3 | 679 | |||
AK173246.1 | - | 6 | 1593 | |||
BC031601.1 | - | 3 | 731 | |||
BC059049.1 | - | 6 | 1692 |