Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbeal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183276.2 | NP_899099.2 | 34 | 5733 | ||
GenBank mRNA | AK129158.1 | - | 10 | 1886 | ||
BC042396.1 | - | 8 | 1228 | |||
BC157956.1 | - | 34 | 5714 | 9007284 | ||
BC172118.1 | - | 33 | 5633 | 9054404 |