Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146229.2 | NP_666341.1 | 8 | 1032 | ||
GenBank mRNA | AK159702.1 | - | 1074 | |||
AK159983.1 | - | 21 | ||||
AK205150.1 | - | 2 | 93 | |||
AK217382.1 | - | 2 | 44 | |||
BC023347.1 | - | 8 | 1032 | 5040044 |