Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290728.1 | NP_001277657.1 | 2 | 184 | ||
NM_177704.3 | NP_808372.1 | 2 | 184 | |||
GenBank mRNA | AB098160.1 | - | 2 | 183 | ||
AB098161.1 | - | 2 | 183 | |||
BC150158.1 | - | 2 | 187 | 40141504 | ||
BC150480.1 | - | 2 | 187 | 40141505 |