Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145628.4 | NP_663603.3 | 9 | 1153 | ||
GenBank mRNA | AK031767.1 | - | 9 | 1132 | ||
AK049053.1 | - | 9 | 1567 | |||
AK220478.1 | - | 8 | 866 | |||
BC005470.1 | - | 7 | 722 | 3496607 |