Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198783.1 | NP_001185712.1 | 12 | 1493 | ||
NM_001198786.1 | NP_001185715.1 | 11 | 1171 | |||
NM_002697.3 | NP_002688.3 | 12 | 1360 | |||
NR_037163.1 | 13 | 1404 | ||||
XM_011509653.2 | XP_011507955.1 | 13 | 1312 | |||
XM_011509654.2 | XP_011507956.1 | 12 | 1273 | |||
XM_011509655.1 | XP_011507957.1 | 13 | 1312 | |||
GenBank mRNA | AK091438.1 | - | 14 | 1788 | ||
AK303201.1 | - | 11 | 1109 | |||
AK308004.1 | - | 13 | 1399 | |||
AY113189.1 | - | 12 | 1312 | |||
BC001664.2 | - | 12 | 1276 | 2966289 | ||
BC003571.1 | - | 12 | 1276 | 3544378 | ||
BC007388.2 | - | 8 | 946 | |||
BC052274.1 | - | 13 | 1384 | 4622256 | ||
KT438684.1 | - | 14 | 1471 | |||
X13403.1 | - | 12 | 1281 |