Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003717.1 | NP_001003717.1 | 7 | 950 | ||
NM_175489.3 | NP_780698.2 | 8 | 987 | |||
GenBank mRNA | AK050244.1 | - | 7 | 951 | ||
BC141383.1 | - | 8 | 686 | 9055996 |