Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029856.2 | NP_001025027.1 | 2 | 808 | ||
GenBank mRNA | AK032814.1 | - | 2 | 746 | ||
AK048355.1 | - | 2 | 770 | |||
AK053857.1 | - | 2 | 798 | |||
AK085211.1 | - | 2 | 799 | |||
AK165945.1 | - | 2 | 775 | |||
AY557610.1 | - | 2 | 799 | |||
BC038279.1 | - | 2 | 816 | |||
BC141180.1 | - | 2 | 466 | 9055792 |