Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030291.1 | NP_001025462.1 | 1, 2 | 270 | ||
GenBank mRNA | AK144474.1 | - | 1, 2 | 273 | ||
AK167678.1 | - | 1, 2 | 273 | |||
BC141245.1 | - | 1, 2 | 355 | 9055857 | ||
BC171967.1 | - | 1, 2 | 355 | 9054253 |