Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf2bpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145836.2 | NP_665835.1 | 1 | 1009 | ||
GenBank mRNA | AF525300.1 | - | 1 | 1006 | ||
AK032366.1 | - | 1 | 1014 | |||
AK032622.1 | - | 1 | 1013 | |||
AK082791.1 | - | 1 | 993 | |||
AK084782.1 | - | 1 | 1014 | |||
AK140441.1 | - | 1 | 990 | |||
AK154383.1 | - | 1 | 759 | |||
AK155373.1 | - | 1 | 964 | |||
AK160059.1 | - | 1 | 963 | |||
AK171043.1 | - | 1 | 964 | |||
AK173282.1 | - | 1 | 1009 | |||
BC049246.1 | - | 1 | 124 | |||
BC057128.1 | - | 1 | 446 | 6810092 | ||
BC063253.1 | - | 1 | 446 | 6810092 |