Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177355.3 | NP_796329.2 | 2 | 886 | ||
GenBank mRNA | AK038575.1 | - | 3 | 977 | ||
AK044972.1 | - | 2 | 733 | |||
BC137637.1 | - | 2 | 741 | 8860656 | ||
BC137639.1 | - | 2 | 741 | 8860658 |