Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177719.4 | NP_808387.2 | 2 | 808 | ||
GenBank mRNA | AK077016.1 | - | 2 | 809 | ||
BC120896.1 | - | 1 | 1005 | 40061539 | ||
BC120897.1 | - | 1 | 1005 | 40061540 |