Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111094.1 | NP_001104564.1 | 4 | 829 | ||
GenBank mRNA | BC052925.1 | - | 4 | 807 | 6398253 |