Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166493.1 | NP_001159965.1 | 12, 13 | 1800 | ||
NM_207246.4 | NP_997129.1 | 13, 14 | 1792 | |||
GenBank mRNA | AB154824.1 | - | 11, 12 | 1487 | ||
AK173041.1 | - | 12, 13 | 1685 | |||
BC066069.1 | - | 11, 12 | 1430 | 6851971 | ||
BC066127.1 | - | 13, 14 | 1717 | 30280332 |