Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mex3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039214.4 | NP_001034303.3 | 2 | 1266 | ||
GenBank mRNA | BC035207.1 | - | 2 | 381 | ||
BC125427.1 | - | 2 | 548 | 40129942 | ||
KC456334.1 | - | 3 | 327 | |||
KC456335.1 | - | 3 | 307 | |||
KC456336.1 | - | 3 | 417 | |||
KC456337.1 | - | 3 | 417 | |||
KC456338.1 | - | 3 | 351 | |||
KC456339.1 | - | 3 | 414 | |||
KC456340.1 | - | 3 | 295 | |||
KC456341.1 | - | 3 | 376 | |||
KC456342.1 | - | 2 | 513 |