Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPOX SIRNA Assays ›
Gene Symbol:
PPOX
Gene Name:
protoporphyrinogen oxidase
Gene Aliases:
PPO, V290M, VP
Chromosome Location:
Chr.1: 161165728 - 161178277 on Build GRCh38
UniGene ID:
Hs.517373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10929
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPOX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000309.3
NP_000300.1
11
1391
NM_001122764.1
NP_001116236.1
11
1452
XM_005245291.3
XP_005245348.2
11
1430
XM_005245295.3
XP_005245352.2
10
1278
XM_006711402.2
XP_006711465.2
11
1397
XM_006711403.2
XP_006711466.2
11
1386
XM_006711404.3
XP_006711467.2
11
1334
XM_006711406.2
XP_006711469.2
11
1307
XM_011509663.2
XP_011507965.1
11
1517
XM_011509664.1
XP_011507966.1
11
1288
XM_011509665.2
XP_011507967.1
11
1515
XM_011509666.2
XP_011507968.1
10
1411
XM_011509667.2
XP_011507969.1
11
1458
XM_011509668.2
XP_011507970.1
11
1258
XM_011509669.1
XP_011507971.1
11
1271
XM_011509670.2
XP_011507972.1
10
1406
XM_011509671.1
XP_011507973.1
11
1288
XM_011509672.2
XP_011507974.1
10
1397
XM_011509673.2
XP_011507975.1
9
1254
XM_011509674.2
XP_011507976.1
8
1232
XM_011509675.2
XP_011507977.1
10
1339
XM_011509676.1
XP_011507978.1
10
1602
XM_011509677.1
XP_011507979.1
10
1535
XM_011509678.1
XP_011507980.1
11
1495
XM_011509679.1
XP_011507981.1
11
1303
XM_011509681.1
XP_011507983.1
10
1220
XM_011509682.1
XP_011507984.1
10
1230
XM_017001559.1
XP_016857048.1
10
1403
XM_017001560.1
XP_016857049.1
10
1319
XM_017001561.1
XP_016857050.1
10
1923
XM_017001562.1
XP_016857051.1
11
1376
XM_017001563.1
XP_016857052.1
11
1474
XM_017001564.1
XP_016857053.1
7
992
XM_017001565.1
XP_016857054.1
11
1376
XM_017001566.1
XP_016857055.1
11
1485
XM_017001567.1
XP_016857056.1
10
1278
XM_017001568.1
XP_016857057.1
11
1466
XM_017001569.1
XP_016857058.1
10
1220
XM_017001570.1
XP_016857059.1
8
1021
XM_017001571.1
XP_016857060.1
4
644
GenBank mRNA
AK094855.1
-
10
1224
AK298912.1
-
5
567
AK302298.1
-
10
1262
BC002357.2
-
11
1372
2821983
D38537.1
-
11
1439
U26446.1
-
11
1353
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00609392_m1
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
response to drug
oxidation-reduction process
oxygen-dependent protoporphyrinogen oxidase activity
flavin adenine dinucleotide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0