Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002709.2 | NP_002700.1 | 6 | 1053 | ||
NM_206876.1 | NP_996759.1 | 7 | 848 | |||
GenBank mRNA | AF092905.1 | - | 8 | 826 | ||
AK295957.1 | - | 6 | 711 | |||
AK312329.1 | - | 6 | 990 | |||
AK316532.1 | - | 6 | 674 | |||
BC002697.2 | - | 6 | 769 | 3609386 | ||
BC012045.1 | - | 6 | 769 | 3631338 | ||
BT019744.1 | - | 6 | 688 | |||
BX647970.1 | - | 7 | 887 | |||
CR542263.1 | - | 6 | 688 | |||
CR542285.1 | - | 6 | 688 | |||
EU668339.1 | - | 6 | 769 | |||
X80910.1 | - | 6 | 946 |