Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172844.2 | NP_766432.1 | 4 | 399 | ||
GenBank mRNA | AK029219.1 | - | 4 | 400 | ||
AK161224.1 | - | 4 | 399 | |||
BC111885.1 | - | 2 | 128 | 40060432 | ||
BC112405.1 | - | 4 | 389 | 40060428 |