Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dytn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081658.1 | NP_001075127.1 | 14 | 1773 | ||
GenBank mRNA | DQ443727.1 | - | 14 | 1773 |