Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101479.1 | NP_001094949.1 | 2 | 533 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB660261.1 | - | 449 | Chr. - - [-] on Build |