Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172295.4 | NP_758499.3 | 4 | 994 | ||
GenBank mRNA | AK029040.1 | - | 4 | 733 | ||
AK161057.1 | - | 4 | 911 | |||
AK166928.1 | - | 744 | ||||
AK172389.1 | - | 4 | 943 | |||
BC037703.1 | - | 4 | 931 | 4166596 |