Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206994.1 | NP_001193923.1 | 5 | 472 | ||
NM_001206995.1 | NP_001193924.1 | 5 | 576 | |||
NM_001206996.1 | NP_001193925.1 | 5 | 626 | |||
NM_020416.3 | NP_065149.2 | 4 | 764 | |||
NM_181876.2 | NP_870991.1 | 4 | 418 | |||
XM_011513495.1 | XP_011511797.1 | 4 | 513 | |||
XM_011513496.1 | XP_011511798.1 | 4 | 528 | |||
GenBank mRNA | AB209183.1 | - | 3 | 4040 | ||
AF086924.1 | - | 4 | 448 | |||
AF145027.1 | - | 4 | 611 | |||
AK093115.1 | - | 4 | 418 | |||
AK296474.1 | - | 5 | 472 | |||
AK316003.1 | - | 6 | 586 | |||
BC032954.2 | - | 5 | 626 | 4823280 |