Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159963.1 | NP_001153435.1 | 5 | 760 | ||
NM_178729.4 | NP_848844.1 | 5 | 760 | |||
GenBank mRNA | AK029957.1 | - | 5 | 708 | ||
AK041497.1 | - | 5 | 732 | |||
AK054009.1 | - | 5 | 730 | |||
AK082931.1 | - | 5 | 730 | |||
AK085100.1 | - | 5 | 733 | |||
AK088067.1 | - | 5 | 727 | |||
AK150664.1 | - | 5 | 720 | |||
AK159744.1 | - | 5 | 749 | |||
AK171361.1 | - | 5 | 714 | |||
AK171783.1 | - | 5 | 620 | |||
AK177606.1 | - | 1 | 73 | |||
AK183837.1 | - | 1 | 73 | |||
BC023291.1 | - | 3 | 365 | |||
BC047214.1 | - | 673 | 6466511 | |||
BC113798.1 | - | 5 | 678 | 40062506 |