Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159963.1 | NP_001153435.1 | 9 | 1885 | ||
NM_178729.4 | NP_848844.1 | 9 | 1885 | |||
GenBank mRNA | AK029957.1 | - | 9 | 1833 | ||
AK041497.1 | - | 9 | 1857 | |||
AK082931.1 | - | 9 | 1855 | |||
AK085100.1 | - | 9 | 1858 | |||
AK088067.1 | - | 9 | 1852 | |||
AK150664.1 | - | 9 | 1842 | |||
AK159744.1 | - | 9 | 1874 | |||
AK171783.1 | - | 9 | 1745 | |||
BC023291.1 | - | 7 | 1490 | |||
BC047214.1 | - | 1798 | 6466511 | |||
BC113798.1 | - | 9 | 1803 | 40062506 |