Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153801.3 | NP_722496.2 | 1 | 368 | ||
GenBank mRNA | AF548365.1 | - | 1 | 369 | ||
AK052276.1 | - | 1 | 207 | |||
AK052284.1 | - | 1 | 371 | |||
AK052302.1 | - | 1 | 73 | |||
AK052378.1 | - | 1 | 131 | |||
BC107366.1 | - | 1 | 281 | 40056219 | ||
BC107367.1 | - | 1 | 281 | 40056223 |