Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081146.2 | NP_001074615.1 | 4 | 417 | ||
NM_001134459.1 | NP_001127931.1 | 3 | 496 | |||
NM_001134460.2 | NP_001127932.1 | 3 | 818 | |||
NM_001134461.1 | NP_001127933.1 | 3 | 313 | |||
GenBank mRNA | AI850364.1 | - | 4 | 359 | ||
AK032631.1 | - | 3 | 821 | |||
AK147576.1 | - | 3 | 497 | |||
AK147628.1 | - | 3 | 497 | |||
BC050793.1 | - | 3 | 313 | |||
BC099897.1 | - | 4 | 388 | 30621797 | ||
BC145754.1 | - | 3 | 335 | 40131057 |