Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153589.2 | NP_705817.2 | 4 | 859 | ||
GenBank mRNA | AK044763.1 | - | 4 | 860 | ||
BC033409.1 | - | 3 | 454 | 5357763 | ||
BC060368.1 | - | 3 | 463 | |||
FJ384096.1 | - | 4 | 860 | |||
KC164759.2 | - | 4 | 859 | |||
KC164761.2 | - | 3 | 552 |