Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175649.5 | NP_783580.1 | 6 | 503 | ||
GenBank mRNA | AK142511.1 | - | 564 | |||
AK154666.1 | - | 6 | 614 | |||
AY165628.1 | - | 6 | 503 | |||
BC111108.1 | - | 6 | 491 | |||
BC138819.1 | - | 6 | 552 | 8861841 | ||
BC138820.1 | - | 6 | 553 | 8861842 | ||
BC152357.1 | - | 6 | 499 | |||
BC152358.1 | - | 6 | 503 | 40055748 |