Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081397.1 | NP_001074866.1 | 4 | 730 | ||
GenBank mRNA | AB429291.1 | - | 3 | 404 | ||
AK082350.1 | - | 4 | 466 | |||
BC072580.1 | - | 4 | 742 | |||
BC082311.1 | - | 4 | 598 | |||
BC151049.1 | - | 4 | 684 | 9087962 | ||
BC151051.1 | - | 4 | 685 | 9087964 | ||
BC157966.1 | - | 4 | 685 | 9007290 | ||
BC172122.1 | - | 4 | 685 | 9054408 |