Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039700.2 | NP_001034789.2 | 31, 32 | 6254 | ||
NM_001286454.1 | NP_001273383.1 | 30, 31 | 6224 | |||
NM_181544.2 | NP_853522.2 | 30, 31 | 6224 | |||
GenBank mRNA | AB290926.1 | - | 30, 31 | 6224 | ||
AY164486.1 | - | 30, 31 | 6224 | |||
DQ382344.1 | - | 30, 31 | 6224 | |||
DQ382345.1 | - | 31, 32 | 6254 | |||
DQ382349.1 | - | 12, 13 | 2044 | |||
DQ382350.1 | - | 2044 |