Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039700.2 | NP_001034789.2 | 29 | 5845 | ||
NM_001286454.1 | NP_001273383.1 | 28 | 5815 | |||
NM_181544.2 | NP_853522.2 | 28 | 5815 | |||
GenBank mRNA | AB290926.1 | - | 28 | 5815 | ||
AY164486.1 | - | 28 | 5815 | |||
DQ382344.1 | - | 28 | 5815 | |||
DQ382345.1 | - | 29 | 5845 | |||
DQ382349.1 | - | 10 | 1635 | |||
DQ382350.1 | - | 1635 |