Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172920.4 | NP_766508.3 | 19 | 1849 | ||
GenBank mRNA | AK031291.1 | - | 1 | 929 | ||
AK143845.1 | - | 20 | 1854 | |||
BC031382.1 | - | 12 | 1027 | |||
BC116782.1 | - | 1 | 417 | |||
BC116784.1 | - | 1 | 417 |