Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161431.1 | NP_001154903.1 | 3 | 2050 | ||
NM_198863.2 | NP_942563.2 | 2 | 2026 | |||
GenBank mRNA | AK044761.1 | - | 2 | 2178 | ||
BC111888.1 | - | 2 | 2025 | 40060548 | ||
BC112406.1 | - | 2 | 2025 | 40060552 |