Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc4h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003916.2 | NP_001003916.1 | 1, 2 | 251 | ||
NM_001289696.1 | NP_001276625.1 | 1, 2 | 251 | |||
NM_001289697.1 | NP_001276626.1 | 1, 2 | 251 | |||
GenBank mRNA | AK129302.1 | - | 1, 2 | 125 | ||
AK155445.1 | - | 1, 2 | 251 | |||
AK162005.1 | - | 1, 2 | 148 | |||
BC050850.1 | - | 1, 2 | 149 | 6508566 | ||
BC079862.1 | - | 1, 2 | 174 | 6807437 | ||
BC094942.1 | - | 1, 2 | 192 | 6435125 | ||
BC119285.1 | - | 1, 2 | 165 | 8734034 | ||
BC119311.1 | - | 1, 2 | 166 | 8734060 | ||
BY200912.1 | - | 1, 2 | 251 |