Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eda2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161432.1 | NP_001154904.1 | 4 | 497 | ||
NM_001161433.1 | NP_001154905.1 | 4 | 452 | |||
NM_175540.5 | NP_780749.3 | 4 | 452 | |||
GenBank mRNA | AK034909.1 | - | 4 | 416 | ||
AK049134.1 | - | 4 | 459 | |||
AK080072.1 | - | 4 | 416 | |||
BC106845.1 | - | 4 | 438 | 40045945 |