Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mum1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164630.1 | NP_001158102.1 | 7 | 2341 | ||
NM_001164631.1 | NP_001158103.1 | 4 | 1923 | |||
NM_001164632.1 | NP_001158104.1 | 5 | 2084 | |||
NM_001164633.1 | NP_001158105.1 | 5 | 2023 | |||
NM_175541.3 | NP_780750.2 | 6 | 2264 | |||
GenBank mRNA | AK035086.1 | - | 1 | 1092 | ||
BC053077.1 | - | 1 | 231 | |||
BC063766.1 | - | 1 | 534 | |||
BC068178.1 | - | 4 | 1942 | |||
BC096534.1 | - | 1 | 1266 | |||
BC120584.1 | - | 1 | 1187 | 40129507 | ||
BC137665.1 | - | 1 | 1187 | 8860684 |