Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177750.6 | NP_808418.1 | 5 | 620 | ||
GenBank mRNA | AK047241.1 | - | 3 | 2340 | ||
BC117734.1 | - | 3 | 276 | 40089821 | ||
BC117735.1 | - | 3 | 276 | 40089822 | ||
BC129957.1 | - | 3 | 276 | 40089827 | ||
BC129958.1 | - | 3 | 276 | 40089829 |