Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003917.4 | NP_001003917.2 | 14 | 2478 | ||
NM_001288612.1 | NP_001275541.1 | 14 | 2481 | |||
NM_001288613.1 | NP_001275542.1 | 14 | 2726 | |||
GenBank mRNA | AM085510.1 | - | 14 | 2391 | ||
BC034139.1 | - | 7 | 1167 | |||
BC056488.1 | - | 14 | 2639 | |||
BC079884.1 | - | 14 | 2367 | 6840050 | ||
BC082307.1 | - | 12 | 2183 |