Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139225.1 | NP_631971.1 | 2 | 102 | ||
GenBank mRNA | AJ437645.1 | - | 2 | 102 |
Gene Symbol: Defb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139221.2 | NP_631967.1 | 2 | 128 | ||
GenBank mRNA | AJ437646.1 | - | 2 | 107 | ||
BC099849.1 | - | 2 | 96 | 40038952 | ||
CJ044726.1 | - | 2 | 129 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK016897.1 | - | 728 | Chr. - - [-] on Build |